Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109535 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD40 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD40 - CD40 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001250.5 | Intron | NP_001241.1 | ||||
NM_001302753.1 | Intron | NP_001289682.1 | ||||
NM_001322421.1 | Intron | NP_001309350.1 | ||||
NM_001322422.1 | Intron | NP_001309351.1 | ||||
NM_152854.3 | Intron | NP_690593.1 | ||||
XM_005260619.3 | Intron | XP_005260676.1 | ||||
XM_011529109.2 | Intron | XP_011527411.1 | ||||
XM_017028135.1 | Intron | XP_016883624.1 | ||||
XM_017028136.1 | Intron | XP_016883625.1 | ||||
XM_017028137.1 | Intron | XP_016883626.1 |
Set Membership: |
HapMap |