Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCACAAATTGATTTTGTCTGCAA[A/G]TTCAATAGCTAAGAAAAGATAGCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606439 MIM: 604923 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ATL1 - atlastin GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127713.1 | Intron | NP_001121185.1 | ||||
NM_015915.4 | Intron | NP_056999.2 | ||||
NM_181598.3 | Intron | NP_853629.2 |
MAP4K5 - mitogen-activated protein kinase kinase kinase kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006575.4 | Intron | NP_006566.2 | ||||
NM_198794.2 | Intron | NP_942089.1 | ||||
XM_006720013.3 | Intron | XP_006720076.1 | ||||
XM_011536377.2 | Intron | XP_011534679.1 | ||||
XM_011536378.2 | Intron | XP_011534680.1 | ||||
XM_011536379.2 | Intron | XP_011534681.1 | ||||
XM_011536380.2 | Intron | XP_011534682.1 |
Set Membership: |
HapMap Validated |