Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615998 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RNF10 - ring finger protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014868.4 | Intron | NP_055683.3 | ||||
XM_005254013.2 | Intron | XP_005254070.1 | ||||
XM_006719717.2 | Intron | XP_006719780.1 | ||||
XM_017020282.1 | Intron | XP_016875771.1 | ||||
XM_017020283.1 | Intron | XP_016875772.1 |