Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACACTAGGAGGGGTTTCCATGGA[A/G]GGCCTTCCCTGAGTTTGCAGTCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616537 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VSTM2L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
VSTM2L - V-set and transmembrane domain containing 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080607.2 | Intron | NP_542174.1 | ||||
XM_011528530.1 | Intron | XP_011526832.1 |
Set Membership: |
Validated |