Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611827 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MRPL14 - mitochondrial ribosomal protein L14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318767.1 | 377 | Silent Mutation | TTC,TTT | F,F 100 | NP_001305696.1 | |
NM_001318768.1 | 377 | Silent Mutation | TTC,TTT | F,F 100 | NP_001305697.1 | |
NM_001318769.1 | 377 | Silent Mutation | TTC,TTT | F,F 111 | NP_001305698.1 | |
NM_001318770.1 | 377 | Silent Mutation | TTC,TTT | F,F 153 | NP_001305699.1 | |
NM_001318771.1 | 377 | Silent Mutation | TTC,TTT | F,F 66 | NP_001305700.1 | |
NM_032111.3 | 377 | Silent Mutation | TTC,TTT | F,F 100 | NP_115487.2 |
Set Membership: |
HapMap |