Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608712 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTPRT - protein tyrosine phosphatase, receptor type T | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007050.5 | 11217 | UTR 3 | NP_008981.4 | |||
NM_133170.3 | 11217 | UTR 3 | NP_573400.3 | |||
XM_017027611.1 | 11217 | UTR 3 | XP_016883100.1 | |||
XM_017027612.1 | 11217 | UTR 3 | XP_016883101.1 | |||
XM_017027613.1 | 11217 | UTR 3 | XP_016883102.1 |
Set Membership: |
HapMap |