Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTGATGGATTCCCCAGTCGGAC[C/G]TGGAGTACTGGGATCTTGGGTGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190090 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SRC - SRC proto-oncogene, non-receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005417.4 | Intron | NP_005408.1 | ||||
NM_198291.2 | Intron | NP_938033.1 | ||||
XM_011529013.2 | Intron | XP_011527315.1 | ||||
XM_017028024.1 | Intron | XP_016883513.1 | ||||
XM_017028025.1 | Intron | XP_016883514.1 | ||||
XM_017028026.1 | Intron | XP_016883515.1 | ||||
XM_017028027.1 | Intron | XP_016883516.1 |