Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOL4L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NOL4L - nucleolar protein 4 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256798.1 | Intron | NP_001243727.1 | ||||
NM_080616.4 | Intron | NP_542183.2 | ||||
XM_005260282.4 | Intron | XP_005260339.1 | ||||
XM_005260283.4 | Intron | XP_005260340.1 | ||||
XM_005260284.4 | Intron | XP_005260341.1 | ||||
XM_005260285.4 | Intron | XP_005260342.2 | ||||
XM_005260286.4 | Intron | XP_005260343.1 | ||||
XM_005260288.2 | Intron | XP_005260345.1 | ||||
XM_005260289.4 | Intron | XP_005260346.1 | ||||
XM_006723702.3 | Intron | XP_006723765.1 | ||||
XM_006723703.3 | Intron | XP_006723766.1 | ||||
XM_011528562.2 | Intron | XP_011526864.1 | ||||
XM_011528563.1 | Intron | XP_011526865.1 | ||||
XM_017027669.1 | Intron | XP_016883158.1 | ||||
XM_017027670.1 | Intron | XP_016883159.1 |