Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTACTTCCTGCCTGGAACTTGAA[A/T]ATAATGTTTAGAGCTGCTGGAGACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611503 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPL PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CENPL - centromere protein L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127181.2 | Intron | NP_001120653.1 | ||||
NM_001171182.1 | Intron | NP_001164653.1 | ||||
NM_033319.3 | Intron | NP_201576.1 |