Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
76 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605494 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EFCAB7 - EF-hand calcium binding domain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032437.3 | 274 | Silent Mutation | GAC,GAT | D,D 8 | NP_115813.2 | |
XM_005271274.3 | 274 | Silent Mutation | GAC,GAT | D,D 8 | XP_005271331.1 | |
XM_006710976.3 | 274 | Silent Mutation | GAC,GAT | D,D 21 | XP_006711039.1 | |
XM_006710977.1 | 274 | Silent Mutation | GAC,GAT | D,D 8 | XP_006711040.1 | |
XM_011542301.2 | 274 | Silent Mutation | GAC,GAT | D,D 21 | XP_011540603.1 | |
XM_017002548.1 | 274 | Silent Mutation | GAC,GAT | D,D 21 | XP_016858037.1 |
ITGB3BP - integrin subunit beta 3 binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |