Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAAATCCAGACCAGTCCTAAAGA[A/G]CAGACCATTGCCTCTATGGACAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
65 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf198 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1orf198 - chromosome 1 open reading frame 198 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136494.1 | Intron | NP_001129966.1 | ||||
NM_001136495.1 | Intron | NP_001129967.1 | ||||
NM_032800.2 | Intron | NP_116189.1 | ||||
XM_017002599.1 | Intron | XP_016858088.1 |
Set Membership: |
HapMap |