Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTATGCCTGTAATTAGCTGACA[C/T]GAGGTAAATGTCCAAAGTCGACCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614118 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSHZ2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TSHZ2 - teashirt zinc finger homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193421.1 | Intron | NP_001180350.1 | ||||
NM_173485.5 | Intron | NP_775756.3 | ||||
XM_017027640.1 | Intron | XP_016883129.1 | ||||
XM_017027641.1 | Intron | XP_016883130.1 |
Set Membership: |
Validated |