Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAATTAAAATAAAATTGGTTGCA[A/T]TTTTAAAATAAAAAAAAAGAGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf36 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C4orf36 - chromosome 4 open reading frame 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144645.3 | Intron | NP_653246.1 | ||||
XM_011531610.2 | Intron | XP_011529912.1 | ||||
XM_011531611.2 | Intron | XP_011529913.1 | ||||
XM_011531612.2 | Intron | XP_011529914.1 | ||||
XM_011531613.2 | Intron | XP_011529915.1 | ||||
XM_011531615.2 | Intron | XP_011529917.1 | ||||
XM_017007746.1 | Intron | XP_016863235.1 |