Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCAGTGGTGACAGTTTGTATCT[C/T]GATTTACCTGCTTCCTTTAAAGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159557 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AFF1 - AF4/FMR2 family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166693.2 | Intron | NP_001160165.1 | ||||
NM_001313959.1 | Intron | NP_001300888.1 | ||||
NM_001313960.1 | Intron | NP_001300889.1 | ||||
NM_005935.3 | Intron | NP_005926.1 | ||||
XM_005263007.3 | Intron | XP_005263064.1 | ||||
XM_005263009.3 | Intron | XP_005263066.1 | ||||
XM_005263011.3 | Intron | XP_005263068.1 | ||||
XM_005263013.3 | Intron | XP_005263070.1 | ||||
XM_011531973.2 | Intron | XP_011530275.1 | ||||
XM_017008215.1 | Intron | XP_016863704.1 | ||||
XM_017008216.1 | Intron | XP_016863705.1 |
LOC100506746 - uncharacterized LOC100506746 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |