Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604358 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STAG1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
STAG1 - stromal antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005862.2 | Intron | NP_005853.2 | ||||
XM_011512329.2 | Intron | XP_011510631.1 | ||||
XM_011512331.2 | Intron | XP_011510633.1 | ||||
XM_017005523.1 | Intron | XP_016861012.1 | ||||
XM_017005524.1 | Intron | XP_016861013.1 | ||||
XM_017005525.1 | Intron | XP_016861014.1 |
Set Membership: |
HapMap |