Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCTCTGTCTCCCCTCCCTCCCC[C/T]TCTCCCAACATGCCCCGCCCTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605144 MIM: 123866 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTR1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTR1B - ARP1 actin-related protein 1 homolog B, centractin beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005735.3 | 1424 | UTR 3 | NP_005726.1 | |||
XM_005263854.4 | 1424 | UTR 3 | XP_005263911.1 | |||
XM_017003116.1 | 1424 | UTR 3 | XP_016858605.1 |
COX5B - cytochrome c oxidase subunit 5B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |