Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTGAATGGAGAATGAAGGCAGG[A/G]GTTTTTTCTTTGGCTTAGCTGCTTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 610911 | |||||||||||||||||||||||
Literature Links: |
ARHGAP31 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ARHGAP31 - Rho GTPase activating protein 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020754.3 | Intron | NP_065805.2 | ||||
XM_006713714.3 | Intron | XP_006713777.1 | ||||
XM_017006955.1 | Intron | XP_016862444.1 |