Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTATAATAAATCCCTTAGTTTT[C/T]GCTGTTCATTAAAAAACAAAAAGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607385 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGAT4C PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MGAT4C - MGAT4 family member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013244.3 | 3797 | Intron | NP_037376.2 | |||
XM_005268776.4 | 3797 | Intron | XP_005268833.1 | |||
XM_005268779.3 | 3797 | UTR 3 | XP_005268836.1 | |||
XM_011538150.2 | 3797 | Intron | XP_011536452.1 | |||
XM_011538151.2 | 3797 | Intron | XP_011536453.1 | |||
XM_011538156.2 | 3797 | UTR 3 | XP_011536458.1 | |||
XM_011538157.2 | 3797 | UTR 3 | XP_011536459.1 | |||
XM_017019143.1 | 3797 | Intron | XP_016874632.1 | |||
XM_017019144.1 | 3797 | Intron | XP_016874633.1 | |||
XM_017019145.1 | 3797 | Intron | XP_016874634.1 | |||
XM_017019146.1 | 3797 | Intron | XP_016874635.1 | |||
XM_017019147.1 | 3797 | Intron | XP_016874636.1 | |||
XM_017019148.1 | 3797 | UTR 3 | XP_016874637.1 | |||
XM_017019149.1 | 3797 | UTR 3 | XP_016874638.1 | |||
XM_017019150.1 | 3797 | UTR 3 | XP_016874639.1 |
Set Membership: |
HapMap |