Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171650 MIM: 600811 MIM: 602423 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ACP2 - acid phosphatase 2, lysosomal | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302489.1 | 1666 | UTR 3 | NP_001289418.1 | |||
NM_001302490.1 | 1666 | UTR 3 | NP_001289419.1 | |||
NM_001302491.1 | 1666 | UTR 3 | NP_001289420.1 | |||
NM_001302492.1 | 1666 | UTR 3 | NP_001289421.1 | |||
NM_001610.3 | 1666 | UTR 3 | NP_001601.1 |
DDB2 - damage specific DNA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000107.2 | 1666 | Intron | NP_000098.1 | |||
NM_001300734.1 | 1666 | Intron | NP_001287663.1 |
NR1H3 - nuclear receptor subfamily 1 group H member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |