Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGCCTGGCGGCTCCCCGAAATC[C/T]CCAGCAAATGGCGCGGTGACCTTGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ARHGEF26 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ARHGEF26 - Rho guanine nucleotide exchange factor 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251962.1 | 592 | Silent Mutation | TCC,TCT | S,S 127 | NP_001238891.1 | |
NM_001251963.1 | 592 | Silent Mutation | TCC,TCT | S,S 127 | NP_001238892.1 | |
NM_015595.3 | 592 | Silent Mutation | TCC,TCT | S,S 127 | NP_056410.3 | |
XM_011512672.1 | 592 | Silent Mutation | TCC,TCT | S,S 127 | XP_011510974.1 | |
XM_017006160.1 | 592 | Intron | XP_016861649.1 |
ARHGEF26-AS1 - ARHGEF26 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |