Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTACCAGCTGTTCTTAGCCTTA[C/T]GTCTGGACAATGTAAATAAAACAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100289361 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100289361 - uncharacterized LOC100289361 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAQR9-AS1 - PAQR9 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
U2SURP - U2 snRNP associated SURP domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080415.1 | Intron | NP_001073884.1 | ||||
NM_001320219.1 | Intron | NP_001307148.1 | ||||
NM_001320220.1 | Intron | NP_001307149.1 | ||||
NM_001320222.1 | Intron | NP_001307151.1 | ||||
XM_005247248.4 | Intron | XP_005247305.1 | ||||
XM_017006037.1 | Intron | XP_016861526.1 | ||||
XM_017006038.1 | Intron | XP_016861527.1 | ||||
XM_017006039.1 | Intron | XP_016861528.1 |
Set Membership: |
HapMap |