Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGGAATGGAAACAAATGGGGCT[A/G]TGTCAGATGCCCGGCTGGCATGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf50 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf50 - chromosome 2 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PQLC3 - PQ loop repeat containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282710.1 | Intron | NP_001269639.1 | ||||
NM_001282711.1 | Intron | NP_001269640.1 | ||||
NM_001282712.1 | Intron | NP_001269641.1 | ||||
NM_152391.4 | Intron | NP_689604.1 | ||||
XM_011510315.1 | Intron | XP_011508617.1 |