Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102772 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMPD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AMPD3 - adenosine monophosphate deaminase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000480.2 | 24 | Intron | NP_000471.1 | |||
NM_001025389.1 | 24 | UTR 5 | NP_001020560.1 | |||
NM_001025390.1 | 24 | Intron | NP_001020561.1 | |||
NM_001172430.1 | 24 | Intron | NP_001165901.1 | |||
NM_001172431.1 | 24 | Intron | NP_001165902.1 |
Set Membership: |
HapMap Validated |