Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600326 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DDX6 - DEAD-box helicase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257191.2 | 5715 | Intron | NP_001244120.1 | |||
NM_004397.5 | 5715 | Intron | NP_004388.2 | |||
XM_005271417.3 | 5715 | Intron | XP_005271474.1 | |||
XM_011542644.1 | 5715 | UTR 3 | XP_011540946.1 | |||
XM_011542645.1 | 5715 | UTR 3 | XP_011540947.1 | |||
XM_017017251.1 | 5715 | Intron | XP_016872740.1 |
Set Membership: |
HapMap |