Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD42 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD42 - ankyrin repeat domain 42 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCDC90B - coiled-coil domain containing 90B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286116.1 | Intron | NP_001273045.1 | ||||
NM_001286117.1 | Intron | NP_001273046.1 | ||||
NM_001286118.1 | Intron | NP_001273047.1 | ||||
NM_001286119.1 | Intron | NP_001273048.1 | ||||
NM_001286120.1 | Intron | NP_001273049.1 | ||||
NM_021825.4 | Intron | NP_068597.2 | ||||
XM_005274154.2 | Intron | XP_005274211.1 | ||||
XM_005274156.3 | Intron | XP_005274213.1 | ||||
XM_005274159.3 | Intron | XP_005274216.1 | ||||
XM_017018103.1 | Intron | XP_016873592.1 | ||||
XM_017018104.1 | Intron | XP_016873593.1 |