Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD42 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD42 - ankyrin repeat domain 42 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCDC90B - coiled-coil domain containing 90B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286116.1 | 2376 | UTR 3 | NP_001273045.1 | |||
NM_001286117.1 | 2376 | UTR 3 | NP_001273046.1 | |||
NM_001286118.1 | 2376 | UTR 3 | NP_001273047.1 | |||
NM_001286119.1 | 2376 | UTR 3 | NP_001273048.1 | |||
NM_001286120.1 | 2376 | UTR 3 | NP_001273049.1 | |||
NM_021825.4 | 2376 | UTR 3 | NP_068597.2 | |||
XM_005274154.2 | 2376 | Intron | XP_005274211.1 | |||
XM_005274156.3 | 2376 | UTR 3 | XP_005274213.1 | |||
XM_005274159.3 | 2376 | UTR 3 | XP_005274216.1 | |||
XM_017018103.1 | 2376 | Intron | XP_016873592.1 | |||
XM_017018104.1 | 2376 | UTR 3 | XP_016873593.1 |