Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601746 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HYOU1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HYOU1 - hypoxia up-regulated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130991.2 | 3459 | UTR 3 | NP_001124463.1 | |||
NM_006389.4 | 3459 | UTR 3 | NP_006380.1 | |||
XM_005271392.3 | 3459 | UTR 3 | XP_005271449.1 | |||
XM_005271393.3 | 3459 | UTR 3 | XP_005271450.1 | |||
XM_005271394.3 | 3459 | UTR 3 | XP_005271451.1 | |||
XM_017017095.1 | 3459 | UTR 3 | XP_016872584.1 | |||
XM_017017096.1 | 3459 | UTR 3 | XP_016872585.1 | |||
XM_017017097.1 | 3459 | UTR 3 | XP_016872586.1 |