Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCCAGTACAGCCATCTGTGCCC[A/G]GACCGCGTGCCCCTCTTCCTGCACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRG3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADGRG3 - adhesion G protein-coupled receptor G3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DRC7 - dynein regulatory complex subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289162.1 | 419 | Silent Mutation | CCA,CCG | P,P 110 | NP_001276091.1 | |
NM_001289163.1 | 419 | Silent Mutation | CCA,CCG | P,P 110 | NP_001276092.1 | |
NM_032269.5 | 419 | Silent Mutation | CCA,CCG | P,P 110 | NP_115645.4 | |
XM_011523377.1 | 419 | Silent Mutation | CCA,CCG | P,P 110 | XP_011521679.1 | |
XM_011523378.1 | 419 | Silent Mutation | CCA,CCG | P,P 110 | XP_011521680.1 |
Set Membership: |
HapMap |