Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCGTGTGAAAGAAGCCAAGCGG[A/C]GGCTGCAGCCCGGGCCGCGATGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606461 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TGS1 - trimethylguanosine synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317902.1 | 143 | UTR 5 | NP_001304831.1 | |||
NM_024831.7 | 143 | UTR 5 | NP_079107.6 | |||
XM_005251328.2 | 143 | Intron | XP_005251385.1 | |||
XM_006716485.2 | 143 | Intron | XP_006716548.1 | |||
XM_017014004.1 | 143 | Intron | XP_016869493.1 |
TMEM68 - transmembrane protein 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286657.1 | 143 | Intron | NP_001273586.1 | |||
NM_001286660.1 | 143 | UTR 5 | NP_001273589.1 | |||
NM_001286661.1 | 143 | UTR 5 | NP_001273590.1 | |||
NM_152417.2 | 143 | UTR 5 | NP_689630.1 | |||
XM_005251150.1 | 143 | UTR 5 | XP_005251207.1 | |||
XM_011517454.1 | 143 | UTR 5 | XP_011515756.1 | |||
XM_011517455.2 | 143 | UTR 5 | XP_011515757.1 | |||
XM_017013040.1 | 143 | UTR 5 | XP_016868529.1 |
Set Membership: |
HapMap |