Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCACCGCCTTAAAGAGCCTGTGT[A/G]AAATGGACTTGAGTAAAATACAGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609182 MIM: 610160 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC35D2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SLC35D2 - solute carrier family 35 member D2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF367 - zinc finger protein 367 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153695.3 | Intron | NP_710162.1 | ||||
XM_011518345.2 | Intron | XP_011516647.1 |
Set Membership: |
HapMap |