Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616520 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHCYL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AHCYL2 - adenosylhomocysteinase like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STRIP2 - striatin interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134336.1 | Intron | NP_001127808.1 | ||||
NM_020704.2 | Intron | NP_065755.1 | ||||
XM_006716069.3 | Intron | XP_006716132.1 | ||||
XM_011516432.2 | Intron | XP_011514734.1 | ||||
XM_011516435.1 | Intron | XP_011514737.1 | ||||
XM_017012470.1 | Intron | XP_016867959.1 | ||||
XM_017012471.1 | Intron | XP_016867960.1 | ||||
XM_017012472.1 | Intron | XP_016867961.1 |