Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTACATACTGTATGATTCCAACC[A/G]TATTACATTTTGGAAAAGGCAAAAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601130 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP2C9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYP2C9 - cytochrome P450 family 2 subfamily C member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000771.3 | Intron | NP_000762.2 | ||||
XM_017015758.1 | Intron | XP_016871247.1 |
Set Membership: |
HapMap |