Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTTTCCTCAGACCCATCTGGGA[C/T]GGGGTCTGAAATGCCACTTTTGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608501 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00994 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LINC00994 - long intergenic non-protein coding RNA 994 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRICKLE2 - prickle planar cell polarity protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198859.3 | 4608 | UTR 3 | NP_942559.1 | |||
XM_011533432.2 | 4608 | UTR 3 | XP_011531734.1 | |||
XM_011533433.2 | 4608 | UTR 3 | XP_011531735.1 | |||
XM_011533434.2 | 4608 | UTR 3 | XP_011531736.1 | |||
XM_011533435.2 | 4608 | UTR 3 | XP_011531737.1 | |||
XM_011533436.2 | 4608 | UTR 3 | XP_011531738.1 | |||
XM_011533437.2 | 4608 | UTR 3 | XP_011531739.1 | |||
XM_011533438.2 | 4608 | UTR 3 | XP_011531740.1 | |||
XM_011533440.2 | 4608 | UTR 3 | XP_011531742.1 | |||
XM_017005798.1 | 4608 | UTR 3 | XP_016861287.1 | |||
XM_017005799.1 | 4608 | UTR 3 | XP_016861288.1 |
PRICKLE2-AS1 - PRICKLE2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRICKLE2-AS2 - PRICKLE2 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |