Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600004 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EPHA5 - EPH receptor A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281765.2 | 5192 | UTR 3 | NP_001268694.1 | |||
NM_001281766.2 | 5192 | UTR 3 | NP_001268695.1 | |||
NM_001281767.2 | 5192 | Intron | NP_001268696.1 | |||
NM_001318761.1 | 5192 | UTR 3 | NP_001305690.1 | |||
NM_004439.7 | 5192 | UTR 3 | NP_004430.4 | |||
NM_182472.4 | 5192 | UTR 3 | NP_872272.2 | |||
XM_005265653.3 | 5192 | UTR 3 | XP_005265710.1 | |||
XM_011531735.2 | 5192 | UTR 3 | XP_011530037.1 | |||
XM_017007878.1 | 5192 | UTR 3 | XP_016863367.1 | |||
XM_017007879.1 | 5192 | Intron | XP_016863368.1 | |||
XM_017007880.1 | 5192 | UTR 3 | XP_016863369.1 | |||
XM_017007881.1 | 5192 | Intron | XP_016863370.1 |
Set Membership: |
HapMap |