Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATTGTGTGTTTGGGACCTGCAT[G/A]TCAATACTGGGGTATTTTGGTAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616778 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUSP22 - dual specificity phosphatase 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286555.1 | Intron | NP_001273484.1 | ||||
NM_020185.4 | Intron | NP_064570.1 | ||||
XM_011514757.2 | Intron | XP_011513059.1 | ||||
XM_017011061.1 | Intron | XP_016866550.1 | ||||
XM_017011062.1 | Intron | XP_016866551.1 |