Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCCAATCAGAGTTGAAACTTTG[C/T]GTGGGCTAGCTTAGTTGAAGAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608704 MIM: 610180 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NMRK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NMRK1 - nicotinamide riboside kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OSTF1 - osteoclast stimulating factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012383.4 | Intron | NP_036515.4 | ||||
XM_006717053.3 | Intron | XP_006717116.1 | ||||
XM_006717055.3 | Intron | XP_006717118.1 | ||||
XM_011518524.2 | Intron | XP_011516826.1 | ||||
XM_011518525.2 | Intron | XP_011516827.1 | ||||
XM_017014622.1 | Intron | XP_016870111.1 |