Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114107 MIM: 608736 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP3CC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPP3CC - protein phosphatase 3 catalytic subunit gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243974.1 | 175 | UTR 5 | NP_001230903.1 | |||
NM_001243975.1 | 175 | UTR 5 | NP_001230904.1 | |||
NM_005605.4 | 175 | UTR 5 | NP_005596.2 | |||
XM_017013610.1 | 175 | Intron | XP_016869099.1 | |||
XM_017013611.1 | 175 | UTR 5 | XP_016869100.1 |
SLC39A14 - solute carrier family 39 member 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |