Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609074 MIM: 603002 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXW9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FBXW9 - F-box and WD repeat domain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD135 - small nucleolar RNA, C/D box 135 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD41 - small nucleolar RNA, C/D box 41 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNPO2 - transportin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136195.1 | 3868 | UTR 3 | NP_001129667.1 | |||
NM_001136196.1 | 3868 | UTR 3 | NP_001129668.1 | |||
NM_013433.4 | 3868 | UTR 3 | NP_038461.2 |
Set Membership: |
HapMap Validated |