Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCATGATCTGCACTCAGACGCTC[A/C]CCATCAGAAAGGAACTGTTTTCTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZMAT4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZMAT4 - zinc finger matrin-type 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135731.1 | 1642 | UTR 3 | NP_001129203.1 | |||
NM_024645.2 | 1642 | UTR 3 | NP_078921.1 | |||
XM_017013836.1 | 1642 | Intron | XP_016869325.1 | |||
XM_017013837.1 | 1642 | Intron | XP_016869326.1 | |||
XM_017013838.1 | 1642 | Intron | XP_016869327.1 | |||
XM_017013839.1 | 1642 | Intron | XP_016869328.1 | |||
XM_017013840.1 | 1642 | Intron | XP_016869329.1 |