Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGACAGTCTAATAATCTGTCTC[C/G]CCTTTAAGCCAAGGTTCTTTTAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601274 | ||||||||||||||||||||
Literature Links: |
PTGR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTGR1 - prostaglandin reductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146108.1 | Intron | NP_001139580.1 | ||||
NM_001146109.1 | Intron | NP_001139581.1 | ||||
NM_012212.3 | Intron | NP_036344.2 | ||||
XM_011518394.2 | Intron | XP_011516696.1 | ||||
XM_011518395.2 | Intron | XP_011516697.1 | ||||
XM_017014485.1 | Intron | XP_016869974.1 |
ZNF483 - zinc finger protein 483 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007169.3 | Intron | NP_001007170.1 | ||||
NM_133464.3 | Intron | NP_597721.2 | ||||
XM_011518300.2 | Intron | XP_011516602.1 | ||||
XM_017014337.1 | Intron | XP_016869826.1 | ||||
XM_017014338.1 | Intron | XP_016869827.1 | ||||
XM_017014339.1 | Intron | XP_016869828.1 |