Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180385 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LMO2 - LIM domain only 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142315.1 | 1741 | UTR 3 | NP_001135787.1 | |||
NM_001142316.1 | 1741 | UTR 3 | NP_001135788.1 | |||
NM_005574.3 | 1741 | UTR 3 | NP_005565.2 | |||
XM_005252921.2 | 1741 | UTR 3 | XP_005252978.1 | |||
XM_017017727.1 | 1741 | UTR 3 | XP_016873216.1 | |||
XM_017017728.1 | 1741 | UTR 3 | XP_016873217.1 | |||
XM_017017729.1 | 1741 | UTR 3 | XP_016873218.1 | |||
XM_017017730.1 | 1741 | UTR 3 | XP_016873219.1 | |||
XM_017017731.1 | 1741 | UTR 3 | XP_016873220.1 | |||
XM_017017732.1 | 1741 | UTR 3 | XP_016873221.1 | |||
XM_017017733.1 | 1741 | UTR 3 | XP_016873222.1 |
Set Membership: |
HapMap Validated |