Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCACTCACTCTGGTCTCCCTTCA[G/T]TTCCTTCTGACCTCTCTCCAGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604226 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARPC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARPC4 - actin related protein 2/3 complex subunit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ARPC4-TTLL3 - ARPC4-TTLL3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198793.1 | Intron | NP_001185722.1 |
TTLL3 - tubulin tyrosine ligase like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025930.3 | Intron | NP_001021100.3 |