Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606653 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGR6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LGR6 - leucine rich repeat containing G protein-coupled receptor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017403.1 | Intron | NP_001017403.1 | ||||
NM_001017404.1 | Intron | NP_001017404.1 | ||||
NM_021636.2 | Intron | NP_067649.2 | ||||
XM_005245404.3 | Intron | XP_005245461.1 | ||||
XM_011509838.2 | Intron | XP_011508140.1 | ||||
XM_011509839.2 | Intron | XP_011508141.1 | ||||
XM_011509840.2 | Intron | XP_011508142.1 | ||||
XM_011509841.2 | Intron | XP_011508143.1 | ||||
XM_011509842.2 | Intron | XP_011508144.1 | ||||
XM_011509843.2 | Intron | XP_011508145.1 | ||||
XM_011509844.2 | Intron | XP_011508146.1 | ||||
XM_011509846.2 | Intron | XP_011508148.1 | ||||
XM_017001996.1 | Intron | XP_016857485.1 | ||||
XM_017001997.1 | Intron | XP_016857486.1 | ||||
XM_017001998.1 | Intron | XP_016857487.1 |