Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCTACAATATTCAGAGTAAAAG[C/T]AAATGAAGATTTTAATGCTCCCACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601524 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRB14 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GRB14 - growth factor receptor bound protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303422.1 | Intron | NP_001290351.1 | ||||
NM_004490.2 | Intron | NP_004481.2 | ||||
XM_011511022.2 | Intron | XP_011509324.1 | ||||
XM_017003899.1 | Intron | XP_016859388.1 |
Set Membership: |
HapMap |