Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608500 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRICKLE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PRICKLE1 - prickle planar cell polarity protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144881.1 | 2692 | UTR 3 | NP_001138353.1 | |||
NM_001144882.1 | 2692 | UTR 3 | NP_001138354.1 | |||
NM_001144883.1 | 2692 | UTR 3 | NP_001138355.1 | |||
NM_153026.2 | 2692 | UTR 3 | NP_694571.2 | |||
XM_011537946.1 | 2692 | UTR 3 | XP_011536248.1 | |||
XM_011537947.2 | 2692 | UTR 3 | XP_011536249.1 | |||
XM_017018838.1 | 2692 | UTR 3 | XP_016874327.1 | |||
XM_017018839.1 | 2692 | UTR 3 | XP_016874328.1 | |||
XM_017018840.1 | 2692 | UTR 3 | XP_016874329.1 |
Set Membership: |
HapMap Validated |