Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605230 MIM: 609610 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TP53BP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TP53BP1 - tumor protein p53 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001141979.1 | 5698 | Intron | NP_001135451.1 | |||
NM_001141980.1 | 5698 | Intron | NP_001135452.1 | |||
NM_005657.2 | 5698 | Intron | NP_005648.1 | |||
XM_011521985.1 | 5698 | Intron | XP_011520287.1 | |||
XM_011521986.2 | 5698 | Intron | XP_011520288.1 | |||
XM_017022533.1 | 5698 | Intron | XP_016878022.1 |
TUBGCP4 - tubulin gamma complex associated protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286414.2 | 5698 | Intron | NP_001273343.1 | |||
NM_014444.4 | 5698 | Intron | NP_055259.2 | |||
XM_011521454.2 | 5698 | UTR 3 | XP_011519756.1 | |||
XM_011521455.2 | 5698 | UTR 3 | XP_011519757.1 | |||
XM_017022078.1 | 5698 | UTR 3 | XP_016877567.1 |
Set Membership: |
HapMap Validated |