Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGACTTGCTGTGAGATGTTATGT[G/T]GATCCCCCATGTGATCTTCTGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611831 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL18 - mitochondrial ribosomal protein L18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PNLDC1 - PARN like, ribonuclease domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271862.1 | Intron | NP_001258791.1 | ||||
NM_173516.2 | Intron | NP_775787.1 | ||||
XM_011535491.2 | Intron | XP_011533793.1 | ||||
XM_011535493.2 | Intron | XP_011533795.1 | ||||
XM_017010312.1 | Intron | XP_016865801.1 | ||||
XM_017010313.1 | Intron | XP_016865802.1 | ||||
XM_017010314.1 | Intron | XP_016865803.1 | ||||
XM_017010315.1 | Intron | XP_016865804.1 | ||||
XM_017010316.1 | Intron | XP_016865805.1 |