Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGTATCACTCACAATTCTGCCT[A/C]AAATACTTTGTTTCACACACAAACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMP27 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MMP27 - matrix metallopeptidase 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022122.2 | Intron | NP_071405.2 | ||||
XM_011542948.2 | Intron | XP_011541250.1 | ||||
XM_011542949.2 | Intron | XP_011541251.1 | ||||
XM_011542950.2 | Intron | XP_011541252.1 | ||||
XM_017018120.1 | Intron | XP_016873609.1 |
Set Membership: |
HapMap |