Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGTCCACCATCTGCCCTCCCCA[C/T]CCACCATGGCCTCTAGACCCCTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179410 MIM: 615001 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RDX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RDX - radixin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260492.1 | Intron | NP_001247421.1 | ||||
NM_001260493.1 | Intron | NP_001247422.1 | ||||
NM_001260494.1 | Intron | NP_001247423.1 | ||||
NM_001260495.1 | Intron | NP_001247424.1 | ||||
NM_001260496.1 | Intron | NP_001247425.1 | ||||
NM_002906.3 | Intron | NP_002897.1 |
ZC3H12C - zinc finger CCCH-type containing 12C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |