Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610004 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL25A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
COL25A1 - collagen type XXV alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256074.1 | 2780 | Intron | NP_001243003.1 | |||
NM_032518.2 | 2780 | Intron | NP_115907.2 | |||
NM_198721.3 | 2780 | UTR 3 | NP_942014.1 | |||
XM_011532333.2 | 2780 | UTR 3 | XP_011530635.1 | |||
XM_011532334.2 | 2780 | UTR 3 | XP_011530636.1 | |||
XM_011532335.2 | 2780 | UTR 3 | XP_011530637.1 | |||
XM_011532338.2 | 2780 | UTR 3 | XP_011530640.1 | |||
XM_011532355.2 | 2780 | UTR 3 | XP_011530657.1 | |||
XM_011532356.2 | 2780 | UTR 3 | XP_011530658.1 | |||
XM_011532358.2 | 2780 | UTR 3 | XP_011530660.1 | |||
XM_017008735.1 | 2780 | UTR 3 | XP_016864224.1 | |||
XM_017008736.1 | 2780 | UTR 3 | XP_016864225.1 | |||
XM_017008737.1 | 2780 | UTR 3 | XP_016864226.1 |
Set Membership: |
HapMap Validated |